Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: met4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111703.1 | NP_001105173.1 | 406 | 96 | ||
GenBank mRNA | AY029557.1 | - | 406 | 96 | ||
BT037723.1 | - | 486 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Zea mays |