Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111786.1 | NP_001105256.1 | 469 | 75 | ||
GenBank mRNA | AY560384.1 | - | 469 | 75 | ||
EU956180.1 | - | 449 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Zea mays |