Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111930.1 | NP_001105400.1 | 100 | 78 | ||
GenBank mRNA | U82230.1 | - | 100 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Zea mays |