Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lon1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112433.1 | NP_001105903.1 | 2152 | 75 | ||
GenBank mRNA | U85494.1 | - | 2152 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Protease Zea mays |