Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100193210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001138363.1 | NP_001131835.1 | 502 | 64 | ||
GenBank mRNA | BT035416.1 | - | 502 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Zea mays |