Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100193210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001138363.1 | NP_001131835.1 | 1075 | 100 | ||
GenBank mRNA | BT035416.1 | - | 1075 | 100 |
Gene Symbol: LOC100501015 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BT060777.2 | - | 100 |
Set Membership: |
Amplicon length between 86 and 100 Zea mays |