Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001138792.2 | NP_001132264.2 | 1280 | 100 | ||
GenBank mRNA | BT036036.1 | - | 611 | 100 | ||
BT064122.1 | - | 1426 | 100 | |||
FJ750248.1 | - | 1280 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Zea mays |