Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: thi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112226.1 | NP_001105696.1 | 147 | 92 | ||
GenBank mRNA | BT041396.1 | - | 168 | 92 | ||
U17350.1 | - | 147 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Zea mays |