Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ark | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111737.1 | NP_001105207.1 | 1854 | 85 | ||
GenBank mRNA | AY188755.1 | - | 1854 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase 3' Most Recommended Assay Zea mays |