Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QSOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332143.1 | NP_001321934.1 | 1 - 2 | 538 | 97 | |
NM_101371.5 | NP_172955.1 | 1 - 2 | 588 | 97 | ||
NM_179333.2 | NP_849664.1 | 1 - 2 | 538 | 97 | ||
GenBank mRNA | AY062528.1 | - | 1 - 2 | 329 | 97 | |
BT001218.1 | - | 1 - 2 | 214 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |