Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332166.1 | NP_001319010.1 | 9 - 10 | 1721 | 80 | |
NM_001332167.1 | NP_001322448.1 | 10 - 11 | 1732 | 80 | ||
NM_001332168.1 | NP_001322449.1 | 8 - 9 | 1536 | 80 | ||
NM_101415.4 | NP_172999.1 | 10 - 11 | 1731 | 80 | ||
GenBank mRNA | AY069887.1 | - | 9 - 10 | 1646 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |