Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332173.1 | NP_001323077.1 | 1 - 2 | 474 | 117 | |
NM_101421.3 | NP_173005.1 | 1 - 2 | 474 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |