Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PnsB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101466.3 | NP_563986.1 | 4 - 5 | 1196 | 149 | |
GenBank mRNA | AY057703.1 | - | 4 - 5 | 1043 | 149 | |
AY084966.1 | - | 4 - 5 | 1043 | 149 | ||
AY124875.1 | - | 4 - 5 | 1027 | 149 | ||
KJ139005.1 | - | 4 - 5 | 1027 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |