Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WAKL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101477.2 | NP_173061.2 | 1 - 2 | 598 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |