Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPCP-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101496.3 | NP_173080.1 | 7 - 8 | 762 | 70 | |
GenBank mRNA | BT021096.1 | - | 7 - 8 | 642 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |