Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATKDSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035978.2 | NP_001031055.1 | 2 - 3 | 144 | 89 | |
NM_001084078.1 | NP_001077547.1 | 2 - 3 | 84 | 89 | ||
NM_001198082.1 | NP_001185011.1 | 1 - 2 | 214 | 89 | ||
NM_101500.4 | NP_173084.1 | 1 - 2 | 333 | 89 | ||
GenBank mRNA | AK175407.1 | - | 2 - 3 | 116 | 89 | |
BT010438.1 | - | 1 - 2 | 41 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |