Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OCT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101505.3 | NP_173089.1 | 1 | 1739 | 146 | |
GenBank mRNA | AY078972.1 | - | 1 | 1652 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |