Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332230.1 | NP_001322901.1 | 8 - 9 | 998 | 102 | |
NM_001332231.1 | NP_001322902.1 | 8 - 9 | 998 | 102 | ||
NM_101519.3 | NP_564001.1 | 8 - 9 | 998 | 102 | ||
GenBank mRNA | AF325457.1 | - | 8 - 9 | 953 | 102 | |
AY034895.1 | - | 8 - 9 | 903 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |