Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332273.1 | NP_001321461.1 | 2 - 3 | 351 | 134 | |
NM_101559.2 | NP_173142.2 | 2 - 3 | 351 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |