Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRFL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198094.1 | NP_001185023.1 | 8 - 9 | 1864 | 99 | |
NM_001332306.1 | NP_001321911.1 | 7 - 8 | 1790 | 99 | ||
NM_101608.5 | NP_564025.2 | 7 - 8 | 1853 | 99 | ||
GenBank mRNA | AY080665.1 | - | 7 - 8 | 1804 | 99 | |
AY122966.1 | - | 6 - 7 | 1615 | 99 | ||
AY519537.1 | - | 6 - 7 | 1615 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |