Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101610.3 | NP_173191.2 | 1 - 2 | 429 | 102 | |
GenBank mRNA | AY702665.1 | - | 1 - 2 | 60 | 102 | |
BT032868.1 | - | 1 - 2 | 60 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |