Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101620.3 | NP_173201.2 | 10 - 11 | 1427 | 69 | |
GenBank mRNA | Z28389.1 | - | 1374 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |