Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101620.3 | NP_173201.2 | 24 - 25 | 3147 | 141 | |
GenBank mRNA | Z28389.1 | - | 3094 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |