Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NF-YA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198095.1 | NP_001185024.1 | 3 - 4 | 865 | 73 | |
NM_101621.3 | NP_173202.1 | 3 - 4 | 679 | 73 | ||
NM_202121.2 | NP_973850.1 | 3 - 4 | 665 | 73 | ||
NM_202122.2 | NP_973851.1 | 2 - 3 | 1021 | 73 | ||
GenBank mRNA | AB493463.1 | - | 2 - 3 | 386 | 73 | |
BT033080.1 | - | 2 - 3 | 386 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |