Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATB BETA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101634.4 | NP_564033.1 | 4 - 5 | 801 | 119 | |
NM_179350.4 | NP_849681.1 | 4 - 5 | 805 | 119 | ||
GenBank mRNA | AF370519.1 | - | 4 - 5 | 695 | 119 | |
AK226540.1 | - | 4 - 5 | 745 | 119 | ||
AY045665.1 | - | 4 - 5 | 692 | 119 | ||
BT000645.1 | - | 4 - 5 | 427 | 119 | ||
BT002183.1 | - | 4 - 5 | 427 | 119 | ||
U40161.1 | - | 4 - 5 | 513 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |