Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332324.1 | NP_001323262.1 | 4 - 5 | 940 | 104 | |
NM_101655.3 | NP_564041.2 | 3 - 4 | 1208 | 104 | ||
GenBank mRNA | AB493465.1 | - | 3 - 4 | 421 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |