Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT1G18070 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035986.1 | NP_001031063.1 | 5 - 6 | 350 | 92 | |
NM_001198097.1 | NP_001185026.1 | 5 - 6 | 355 | 92 | ||
NM_101669.3 | NP_173247.1 | 5 - 6 | 399 | 92 | ||
GenBank mRNA | AY120769.1 | - | 5 - 6 | 334 | 92 | |
BT008889.1 | - | 4 - 5 | 226 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |