Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E12A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101672.4 | NP_173250.1 | 3 - 4 | 379 | 67 | |
GenBank mRNA | AF147721.1 | - | 3 - 4 | 347 | 67 | |
AF332406.1 | - | 3 - 4 | 295 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |