Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101702.4 | NP_564051.1 | 10 - 11 | 871 | 121 | |
GenBank mRNA | AK228098.1 | - | 10 - 11 | 810 | 121 | |
BT006474.1 | - | 9 - 10 | 720 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |