Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101702.4 | NP_564051.1 | 9 - 10 | 807 | 84 | |
GenBank mRNA | AK228098.1 | - | 9 - 10 | 746 | 84 | |
BT006474.1 | - | 8 - 9 | 656 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |