Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TET16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101709.2 | NP_173286.2 | 1 - 2 | 448 | 71 | |
GenBank mRNA | AY924674.1 | - | 1 - 2 | 448 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |