Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TET11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101710.3 | NP_564056.1 | 1 - 2 | 655 | 94 | |
GenBank mRNA | DQ446262.1 | - | 1 - 2 | 510 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |