Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332354.1 | NP_001320464.1 | 4 - 5 | 607 | 126 | |
NM_202129.3 | NP_973858.1 | 4 - 5 | 610 | 126 | ||
GenBank mRNA | AY065351.1 | - | 4 - 5 | 601 | 126 | |
AY087385.1 | - | 4 - 5 | 562 | 126 | ||
AY096687.1 | - | 3 - 4 | 464 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |