Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGL65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035989.1 | NP_001031066.1 | 4 - 5 | 263 | 111 | |
NM_001332367.1 | NP_001321506.1 | 5 - 6 | 835 | 111 | ||
NM_101733.6 | NP_173310.2 | 5 - 6 | 740 | 111 | ||
GenBank mRNA | AY143172.1 | - | 4 - 5 | 328 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |