Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGL65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035989.1 | NP_001031066.1 | 5 - 6 | 280 | 124 | |
NM_001332367.1 | NP_001321506.1 | 6 - 7 | 852 | 124 | ||
NM_101733.6 | NP_173310.2 | 6 - 7 | 757 | 124 | ||
GenBank mRNA | AY143172.1 | - | 5 - 6 | 345 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |