Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQD30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035990.2 | NP_001031067.1 | 6 - 7 | 1069 | 127 | |
NM_001332375.1 | NP_001322630.1 | 5 - 6 | 1187 | 127 | ||
NM_101741.4 | NP_173318.2 | 6 - 7 | 1224 | 127 | ||
GenBank mRNA | BT022035.1 | - | 6 - 7 | 756 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |