Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332376.1 | NP_001319038.1 | 3 - 4 | 933 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |