Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160879.1 | NP_001154351.1 | 12 - 13 | 1500 | 69 | |
NM_001332377.1 | NP_001321112.1 | 12 - 13 | 1589 | 69 | ||
NM_101744.7 | NP_173321.4 | 13 - 14 | 1661 | 69 | ||
GenBank mRNA | EU589462.1 | - | 13 - 14 | 1605 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |