Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101808.5 | NP_564087.2 | 1 - 2 | 401 | 93 | |
GenBank mRNA | AK119034.1 | - | 1 - 2 | 358 | 93 | |
BT005657.1 | - | 1 - 2 | 271 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |