Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198109.2 | NP_001185038.1 | 1 | 1150 | 116 | |
GenBank mRNA | AY085135.1 | - | 1 | 1150 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |