Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WOX14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332461.1 | NP_001322715.1 | 1 - 2 | 205 | 109 | |
NM_101922.3 | NP_173493.2 | 1 - 2 | 205 | 109 | ||
GenBank mRNA | AJ441297.1 | - | 1 - 2 | 205 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |