Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332465.1 | NP_001323170.1 | 3 - 4 | 1441 | 81 | |
NM_001332466.1 | NP_001323169.1 | 2 - 3 | 1529 | 81 | ||
NM_101930.4 | NP_564125.2 | 3 - 4 | 1248 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |