Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332471.1 | NP_001319055.1 | 6 - 7 | 2039 | 89 | |
NM_101937.6 | NP_173508.1 | 5 - 6 | 2229 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |