Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUC-L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103785.4 | NP_175322.1 | 12 - 13 | 1270 | 57 | |
GenBank mRNA | BT005793.1 | - | 12 - 13 | 1165 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |