Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126003.3 | NP_569027.2 | 2 - 3 | 900 | 69 | |
NM_203271.2 | NP_975000.1 | 2 - 3 | 900 | 69 | ||
GenBank mRNA | AK316680.1 | - | 2 - 3 | 808 | 69 | |
AY052363.1 | - | 2 - 3 | 802 | 69 | ||
BT001055.1 | - | 2 - 3 | 771 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |