Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126003.3 | NP_569027.2 | 3 - 4 | 1002 | 82 | |
NM_203271.2 | NP_975000.1 | 3 - 4 | 1002 | 82 | ||
GenBank mRNA | AK316680.1 | - | 3 - 4 | 910 | 82 | |
AY052363.1 | - | 3 - 4 | 904 | 82 | ||
BT001055.1 | - | 3 - 4 | 873 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |