Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATRAD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345748.1 | NP_001330043.1 | 10 - 11 | 1827 | 81 | |
NM_126011.2 | NP_201414.1 | 10 - 11 | 1875 | 81 | ||
GenBank mRNA | AB030250.1 | - | 10 - 11 | 1775 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |