Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085336.1 | NP_001078805.1 | 3 - 4 | 647 | 78 | |
NM_126017.5 | NP_201420.1 | 3 - 4 | 652 | 78 | ||
GenBank mRNA | AY072112.1 | - | 3 - 4 | 543 | 78 | |
AY096665.1 | - | 3 - 4 | 465 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |