Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126030.6 | NP_201433.1 | 2 - 3 | 705 | 79 | |
NM_203273.2 | NP_975002.1 | 1 - 2 | 864 | 79 | ||
GenBank mRNA | AY136450.1 | - | 2 - 3 | 487 | 79 | |
BT010367.1 | - | 1 - 2 | 325 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |