Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLP3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126039.4 | NP_569033.1 | 1 - 2 | 562 | 66 | |
GenBank mRNA | AY086393.1 | - | 1 - 2 | 193 | 66 | |
BT003998.1 | - | 1 - 2 | 229 | 66 | ||
BT005683.1 | - | 1 - 2 | 90 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |